Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ p53 Activator III, RITA

Catalog No. 5061491MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-614-91MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-614-91MG Supplier MilliporeSigma™ Supplier No. 5061491MG
Add to Cart
Add to Cart

The p53 Activator III, RITA, also referenced under CAS 213261-59-7, modulates the biological activity of p53. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Description 97% by HPLC
Target Proteins p53
CAS 213261-59-7
Molecular Formula C14H12O3S2
Formula Weight 292.4g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Crystalline Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.