Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ p53 Modulator, CP-31398

Catalog No. 5061665MG Shop All MilliporeSigma Products
Change view
Click to view available options
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-616-65MG 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-616-65MG Supplier MilliporeSigma™ Supplier No. 5061665MG

A cell-permeable styrylquinazoline compound that protects wild-type p53 against heat-induced denaturation and locks newly synthesized mutant p53 in an active conformation.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 90% by HPLC
CAS 259199-65-0
Molecular Formula C22H26N4O·2HCl
Formula Weight 435.4g/mol
Solubility DMSO
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.