Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ p75NTR Signaling Inhibitor, Cell-permeable, TAT-Pep5

Catalog No. 506181500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-618-1500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-618-1500UG Supplier MilliporeSigma™ Supplier No. 506181500UG
Add to Cart
Add to Cart

The p75NTR Signaling Inhibitor, Cell-permeable, TAT-Pep5 controls the biological activity of p75NTR. This small molecule/inhibitor is primarily used for Neuroscience applications.

  • Packaged Under Inert Gas
  • Peptide Sequence: H2N-Tyr-Gly-Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg-Cys12-Phe-Phe-Arg-Gly-Gly-Phe-Phe-Asn-His-Asn-Pro-Arg-Tyr-Cys26-OHdisulfide bond (12-26)
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a trifluoroacetate salt.
Description 95% by HPLC
Target Proteins Blocker of MAG- and Nogo-induced inhibition of neurite outgrowth
Molecular Formula C150H227N57O32S2
Formula Weight 3404.9g/mol
Solubility DMSO
Content And Storage Hygroscopic, Protect from light
Form Lyophilized
Quantity 500 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.