Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Paclitaxel, Semi-Synthetic

Catalog No. 5805565MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58-055-65MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 58-055-65MG Supplier MilliporeSigma™ Supplier No. 5805565MG
Add to Cart
Add to Cart

A semi-synthetic form of Paclitaxel having no contamination by natural taxanes. Also has reduced levels of epimerization products as contaminants.

A semi-synthetic form of Paclitaxel (Cat. No. 580555) having no contamination by natural taxanes. Also has reduced levels of epimerization products as contaminants.
TRUSTED_SUSTAINABILITY

Specifications

Description 97% by HPLC
Target Proteins Promotes assembly of microtubules and inhibits tubulin disassembly.
CAS 33069-62-4
Molecular Formula C47H51NO14
Formula Weight 853.9g/mol
Solubility DMSO or MeOH
Content And Storage Protect from Light, Protect from Moisture
Form Crystalline Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.