Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PAD Inhibitor, Cl-amidine

Catalog No. 50628210MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-628-210MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-628-210MG Supplier MilliporeSigma™ Supplier No. 50628210MG
Add to Cart
Add to Cart

Cl-amidine is a cell-permeable pan PAD inhibitor (IC50 = 0.8, 6.2, and 5.9 μM for PAD1, PAD3, and PAD4, respectively). Inactivates the calcium bound form of PAD4 in an irreversible manner.

A cell-permeable compound that acts as a pan PAD inhibitor (IC50 = 0.8μM, 6.2μM and 5.9μM for PAD1, PAD3, and PAD4, respectively) in enzymatic assays. It preferentially inactivates the calcium bound form of PAD4 in an irreversible manner, and is shown to complete with BAEE for the active site residue of this enzyme. A small but significant reduction in the efficiency of p300GDB-GRIP1 interaction is observed in transiently transfected CV-1 cells treated with Cl-amidine at concentrations between 25μM and 200μM, in vivo . Furthermore, this compound (200μM) together with HDAC inhibitor SAHA (0.4μM) has shown additive effects in inducing p21, GADD45, and PUMA expression and inhibiting cancer cell growth in a p53-dependent manner in U2OS cells.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description >95% by HPLC
Molecular Formula C16H20ClF3N4O4
Formula Weight 424.8g/mol
Solubility DMSO
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.