Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ SB 203580

Catalog No. 5593891MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-938-91MG 1 mg
55-938-95MG 5 mg
55-938-910MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 55-938-91MG Supplier MilliporeSigma™ Supplier No. 5593891MG
Add to Cart
Add to Cart

SB 203580, CAS 152121-47-6, is a highly specific, potent, cell-permeable, selective, reversible, and ATP-competitive inhibitor of p38 MAP kinase (IC50 = 34 nM in vitro, 600 nM in cells).

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Target Proteins P38MAPK
CAS 152121-47-6
Molecular Formula C21H16N3OSF
Formula Weight 377.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Thin Film
Quantity 1 mg
Purity 98% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.