Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PARP Inhibitor XII

Catalog No. 5288225MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-882-25MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-882-25MG Supplier MilliporeSigma™ Supplier No. 5288225MG
Add to Cart
Add to Cart

The PARP Inhibitor XII, also referenced under CAS 489457-67-2, controls the biological activity of PARP. This small molecule/inhibitor is primarily used for Cell Structure applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Light Brown
Description 95% by HPLC
CAS 489457-67-2
Molecular Formula C15H10ClN3O
Formula Weight 283.7g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.