Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PD 169316

Catalog No. 5130301MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-303-01MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 51-303-01MG Supplier MilliporeSigma™ Supplier No. 5130301MG
Add to Cart
Add to Cart

A potent, cell-permeable, reversible, competitive, and selective p38 MAP kinase inhibitor (IC50 = 89 nM).

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Orange-yellow
Description 98% by HPLC
Target Proteins p38 MAP kinase
CAS 152121-53-4
Molecular Formula C20H13FN4O2
Formula Weight 360.3g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.