Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PDE4D Inhibitor, GEBR-7b

Catalog No. 5247485MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-474-85MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-474-85MG Supplier MilliporeSigma™ Supplier No. 5247485MG
Add to Cart
Add to Cart

The PDE4D Inhibitor, GEBR-7b controls the biological activity of PDE4D. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Pale Yellow
Description 95% by HPLC
Molecular Formula C21H30N2O5
Formula Weight 390.47g/mol
Solubility DMSO
Content And Storage Protect from light
Form Semi-solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.