Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PDK1/Akt/Flt Dual Pathway Inhibitor

Catalog No. 5212755MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-127-55MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-127-55MG Supplier MilliporeSigma™ Supplier No. 5212755MG
Add to Cart
Add to Cart

The PDK1/Akt/Flt Dual Pathway Inhibitor, also referenced under CAS 331253-86-2, controls the biological activity of PDK1/Akt/Flt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 98% by HPLC (sum of two isomers)
Target Proteins Both PDK1 and Akt activities
CAS 331253-86-2
Molecular Formula C10H4N6O
Formula Weight 224.2g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.