Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Pepstatin A Methyl Ester

Catalog No. 5164851MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-648-51MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 51-648-51MG Supplier MilliporeSigma™ Supplier No. 5164851MG
Add to Cart
Add to Cart

A cell-permeable methyl ester derivative of Pepstatin A that acts as a potent, non-competitive, transition-state analog inhibitor of γ-secretase.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Description 95% by HPLC
Target Proteins γ-secretase
Molecular Formula C35H65N5O9
Formula Weight 699.9g/mol
Solubility DMSO
Form Lyophilized
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.