Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PERK Inhibitor II, GSK2656157

Catalog No. 5046510001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-465-10001 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-465-10001 Supplier MilliporeSigma™ Supplier No. 5046510001
Add to Cart
Add to Cart

PERK Inhibitor II, GSK2656157, CAS 1337532-29-2 is a cell-permeable, potent, ATP-competitive inhibitor of PERK/EIF2AK3 (IC50 = 0.9nM).

A cell-permeable pyrrolopyrimidinamine compound that acts as a potent, ATP-competitive EIF2AK3/PERK inhibitor (IC50 = 0.9nM; [ATP] = 10μM), displaying ≥511-fold selectivity over HRI/EIF2AK1, BRK, PKR/EIF2AK2, and MEKK2 (IC50 = 460, 822, 905, and 954 nM, respectively) and much reduced or little activity toward more than 300 other kinases. Potently prevents ER stress-induced enhancement of PERK and eIF2α phosphorylation as well as ATF4 and CHOP upregulation in human BxPC3 and murine LL/2 cultures in vitro (IC50 ≤30nM; 1 h drug preincubation prior to 6 h 1μM Thapsigargin treatment, Cat. No. 586005) and exhibits antitumor efficacy against 3 pancreas and 1 multiple myeloma xenografts in mice (54-110% tumor growth inhibition; 150mg/kg/12 h via p.o.) in vivo . More selective than GSK2606414 (Cat. No. 516535).
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Target Proteins PERK
Molecular Formula C23H21FN6O
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.