Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PFKFB3 Inhibitor II, PFK15

Catalog No. 5310050001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-100-50001 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-100-50001 Supplier MilliporeSigma™ Supplier No. 5310050001
Add to Cart
Add to Cart

A cell-permeable, bioavailable and more potent quinolinyl analog of PFKFB3 Inhibitor, 3PO (Cat

A cell-permeable, bioavailable and more potent quinolinyl analog of PFKFB3 Inhibitor, 3PO (Cat. No. 525330) that inhibits the activity of 6-Phosphofructo-2-kinase (PFKFB3) by reversibly docking to its substrate binding domain (IC50 = 207nM compared to 22.9μM for 3PO). Shown to effectively reduce fructose 2,6-bisphosphate, glucose uptake, and intracellular ATP levels. Does neither affect the activity of 96 other kinases in any significant manner (at ∽10μM) nor PFK-1, hexokinase, phosphoglucoisomerase, or PFKFB4 even at 100μM. Induces apoptosis and reduces proliferation in transformed cells (mean GI50 of ∽3μM in the NCI-60 cancer cell panel). Displays adequate pharmacokinetic properties to suppress glucose uptake and the growth of Lewis lung carcinoma is syngenic mice and block the growth of human tumor xenografts in athymic mice (∽25mg/kg, i.p. administered every 3rd day).

Please note that the molecular weight for this compound is batch-specific due to variable water content.
TRUSTED_SUSTAINABILITY

Specifications

Color Light Beige
Description 97% by HPLC
Target Proteins PFKFB3
Molecular Formula C17H12N2O
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.