Learn More
Description
EMD Millipore Calbiochem™ PHM-L LIPOSORB™ Absorbent has low solubility good chemical stability.
- Antibody content coagulation factors remain intact
- Improved serum or plasma stability
- Low solubility in most solvents has good chemical stability in acids bases
- Resistant to enzymatic degradation to activation of coagulation factors
- Heat stable, sterilized by autoclaving (120°C) without altering its binding capacity
- High capacity with no volume limitation is simple to use without expensive apparatus
- One gram of absorbent is sufficient to remove lipids from 25mL of serum or ascites
Recommended Storage
15° to 30°C
Specifications
Specifications
| Description | PHM-L LIPOSORB Absorbent |
| Formulation | Polyhydroxymethylene substituted by fat oxethylized alcohol. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.