Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Phosphatase Inhibitor Cocktail Set I

Catalog No. 5246241SET Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-462-41SET 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-462-41SET Supplier MilliporeSigma™ Supplier No. 5246241SET
Add to Cart
Add to Cart

Phosphatase Inhibitor Cocktail Set I contains three inhibitors that will inhibit alkaline phosphatases as well as serine/threonine protein phosphatases such as PP1 and PP2A.

A cocktail of three inhibitors that will inhibit alkaline phosphatases as well as serine/threonine protein phosphatases such as PP1 and PP2A. This product is provided as a set of five 1mL vials. Each vial contains 1mL phosphatase inhibitor cocktail solubilized in DMSO with the following components: Bromotetramisole oxalate (2.5 mM), Cantharidin (Cat. No. 210155; 500μM), and Microcystin-LR (Cat. No. 475815; 500 nM).
TRUSTED_SUSTAINABILITY

Specifications

Formulation In DMSO.
Target Proteins Alkaline phosphatases as well as serine/threonine protein phosphatases such as PP1 and PP2A
Form Liquid
Quantity 1 Set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.