Learn More
MilliporeSigma™ Calbiochem™ PhosphoDetect™ Phosphoserine Detection Kit
Catalog No. 5252821KIT Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
52-528-21KIT 1 Kit
1 options
Catalog No. 52-528-21KIT Supplier MilliporeSigma™ Supplier No. 5252821KIT
Add to Cart
Add to Cart
Provides set of four different monoclonal antibodies specific for serine phosphorylation sites.
EMD Millipore Calbiochem™ PhosphoDetect™ Phosphoserine Detection Kit recognizes serine phosphorylated proteins.
  • Recognizes serine phosphorylated proteins in Arabidopsis, bacteria, chicken, human, maize, mouse, rat, tobacco, Xenopus, yeast, zebrafish
  • Phosphorylation patterns in cell extracts may differ when probed with different antibodies
  • Packaging: Glass Bottle
Recommended Storage
−20°C
TRUSTED_SUSTAINABILITY
Specifications
Description PhosphoDetect Phosphoserine Detection Kit
Formulation 25μg ea. antibody lyophilized from 2X PBS, PEG, sucrose and 200μL control phosphoproteins lyophilized 20mM Na2HPO4, 0.1% SDS.
Kit Contents 25μg of ea. of the Phosphoserine Antibodies including clones 1C8, 4A3, 4A9, and 16B4, Rabbit Muscle Positive Control, and data sheet
Quantity 1 Kit