Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Phospholipase C Activator, m-3M3FBS

Catalog No. 52518510MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-518-510MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-518-510MG Supplier MilliporeSigma™ Supplier No. 52518510MG
Add to Cart
Add to Cart

The Phospholipase C Activator, m-3M3FBS, also referenced under CAS 200933-14-8, controls the biological activity of Phospholipase C. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 95% by HPLC
Target Proteins Phospholipase C (PLC)
CAS 200933-14-8
Molecular Formula C16H16F3NO2S
Formula Weight 343.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.