Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Phosphoramidon, Disodium Salt

Catalog No. 5252765MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-527-65MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-527-65MG Supplier MilliporeSigma™ Supplier No. 5252765MG
Add to Cart
Add to Cart

Inhibits some metalloendopeptidases.

Inhibits some metalloendopeptidases. Highly specific inhibitor of thermolysin. Inhibits the conversion of big endothelin-1 to endothelin (IC50 = 4.6μM).
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description Single spot by TLC
Target Proteins thermolysin
Molecular Formula C23H32N3O10P·2Na
Formula Weight 587.5g/mol
Solubility DMSO, MeOH, or H2O
Content And Storage Hygroscopic, Protect from light
Form Lyophilized
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.