Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PI 3-Kα Inhibitor IV

Catalog No. 5281115MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-811-15MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-811-15MG Supplier MilliporeSigma™ Supplier No. 5281115MG
Add to Cart
Add to Cart

The PI 3-Kα Inhibitor IV, also referenced under CAS 1188890-32-5, controls the biological activity of PI 3-Kα. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 95% by HPLC
Target Proteins PI 3-Kα
CAS 1188890-32-5
Molecular Formula C16H15N3O2S·2HCl
Formula Weight 386.3g/mol
Solubility DMSO
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.