Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PI 3-Kγ Inhibitor

Catalog No. 5281065MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-810-65MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-810-65MG Supplier MilliporeSigma™ Supplier No. 5281065MG
Add to Cart
Add to Cart

The PI 3-Kγ Inhibitor, also referenced under CAS 648450-29-7, controls the biological activity of PI 3-Kγ. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Pink
Description 90% by HPLC
Target Proteins Phosphatidylinositol 3-kinase γ (PI 3-Kγ)
CAS 648450-29-7
Molecular Formula C12H7N3O2S
Formula Weight 257.3g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.