Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PI3Kδ Inhibitor, SW30

Catalog No. 5265595MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-655-95MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-655-95MG Supplier MilliporeSigma™ Supplier No. 5265595MG
Add to Cart
Add to Cart

The PI3Kδ Inhibitor, SW30 controls the biological activity of PI3Kδ. This small molecule/inhibitor is primarily used for Membrane applications.

A cell permeable, ATP-competitive and selective tolyl quinazolinone inhibitor of PI 3-Kδ (IC50 = 7 nM, 1300 nM, 85000 nM, and 740nM for PI 3-Kδ PI 3-Kγ PI 3-Kα and PI 3-Kβ, respectively) in an enzymatic assay with an EC50 = 16nM for PI 3-Kδ and 2.7μM for PI 3-Kγ in a cell based assay. It elicits an inflammatory marker suppression effect in co-cultures of HUVEC and PBMCs under superantigen stimulated conditions. This compound, at concentrations between 370nM and 10μM, leads to a dose-dependent decrease in E-selectin expression. Furthermore, it is not cytotoxic to HUVEC cells.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description >95% by HPLC
Molecular Formula C25H21N7O2
Formula Weight 451.48g/mol
Solubility DMSO
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.