Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Piceatannol

Catalog No. 5279481MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-794-81MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-794-81MG Supplier MilliporeSigma™ Supplier No. 5279481MG
Add to Cart
Add to Cart

A cell-permeable, substrate competitive and reversible plant metabolite that inhibits the activity of rat liver protein kinase A catalytic subunit (IC50 = 3μM), PKC (IC50 = 8μM), MLCK (IC50 = 12μM).

A cell-permeable, substrate competitive and reversible plant metabolite that inhibits the activity of rat liver protein kinase A catalytic subunit (IC50 = 3μM), PKC (IC50 = 8μM), MLCK (IC50 = 12μM). In RBL-2H3 cells, the selective inhibition of Syk by piceatannol results in inhibition of FceR1-mediated signaling. Also inhibits wheat embryo Ca2+-dependent protein kinase (CDPK) (IC50 = 19μM), and p72 syk (IC50 = 10μM), a non-receptor tyrosine kinase, relative to Lyn in isolated enzyme preparations. Also possesses anti-leukemic properties. Reported to activate sirtuins and promote the survival of eukaryotic cells.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 95% by HPLC
Target Proteins Activity of rat liver protein kinase A catalytic subunit
CAS 10083-24-6
Molecular Formula C14H12O4
Formula Weight 244.2g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Crystalline Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.