Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PICK1 PDZ Domain Inhibitor, FSC231

Catalog No. 52953110MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-953-110MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-953-110MG Supplier MilliporeSigma™ Supplier No. 52953110MG
Add to Cart
Add to Cart

The PICK1 PDZ Domain Inhibitor, FSC231 controls the biological activity of PICK1. This small molecule/inhibitor is primarily used for Neuroscience applications.

A cell-permeable acryloylcarbamate compound that selectively targets the PDZ (PSD-95/Discs-large/ZO-1 homology) domain of PICK1 (protein interacting with C kinase 1), but not those of PSD-95 (postsynaptic density protein 95) and GRIP1 (glutamate receptor interacting protein 1), effectively competing against dopamine transporter/DAT ( K i ∽10μM), GluR2 ( K i ∽10μM), and mRluR7a, c-terminus binding to PICK1 PDZ. Shown to accelerate internalized GluR2 surface recycling and suppress both long-term depression and potentiation in rat and murine hippocampal neurons.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Molecular Formula C13H10Cl2N2O3
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.