Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Picrotoxin, Anamirta cocculin

Catalog No. 528105100MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-810-5100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-810-5100MG Supplier MilliporeSigma™ Supplier No. 528105100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A potent antagonist of GABA receptors (IC50 = 240 nM) at synapses in the central nervous system.

A potent antagonist of GABA receptors (IC50 = 240 nM) at synapses in the central nervous system. Inhibits 35S-TBPS binding to the Cl channel (IC50 = 1.7μM). Picrotoxin is a mixture of one mole of picrotoxinin and one mole of picrotin.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by TLC
Target Proteins GABA receptors
CAS 124-87-8
Molecular Formula C15H18O7·C15H16O6
Formula Weight 602.6g/mol
Solubility EtOH or basic aqueous solutions
Form Crystalline Solid
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.