Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Pifithrin-μ

Catalog No. 50615510MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-615-510MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-615-510MG Supplier MilliporeSigma™ Supplier No. 50615510MG
Add to Cart
Add to Cart

Pifithrin-μ, CAS 64984-31-2, is a cell-permeable inhibitor of p53 interaction with Bcl-xL and Bcl-2. Inhibits p53 translocation to mitochondria without affecting the transactivation function of p53.

A cell-permeable sulfonamide that blocks p53 interaction with Bcl-xL and Bcl-2 proteins and selectively inhibits p53 translocation to mitochondria without affecting the transactivation function of p53. Effectively protects against γ radiation-induced cell death in vitro and animal lethality in vivo . Because Pifithrin-μ targets only the mitochondrial branch of the p53 pathway without affecting the important transcriptional functions of p53, it is superior to Pifithrin-α (Cat. No. 506132) in in vivo studies. Shown to selectively interact with inducible HSP70 and disrupt its functions.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 95% by HPLC
Target Proteins p53 interaction with Bcl-xL and Bcl-2 proteins
CAS 64984-31-2
Molecular Formula C8H7NO2S
Formula Weight 181.2g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.