Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PIH

Catalog No. 52811050MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-811-050MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-811-050MG Supplier MilliporeSigma™ Supplier No. 52811050MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cell-permeable, non-toxic tridendate iron (Fe3+) chelator of aroyl hydrazone class

  • Highly effective in mobilizing Fe3+ from cells and preventing its uptake from transferrin, even at low concentrations
  • Exhibits similar specificity and binding affinity as Deferoxamine Mesylate but has greater access to mitochondrial iron
  • Also useful for the management of iron overload disease in experimental models
  • Inhibits the induction of heme containing indoleamine 2,3-dioxygenase activity
TRUSTED_SUSTAINABILITY

Specifications

Membrane Permeable Y
Purity ≥99% by HPLC
CAS 737-86-0
Content And Storage 2°C to 8°C
Form Solid
Quantity 50 mg
Synonym Pyridoxal isonicotinoyl hydrazone, Iron Chelator III
Solubility DMSO, Slightly Acidic Medium
Product Type Calbiochem PIH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.