Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Pimaricin, Streptomyces chattanoogensis

Catalog No. 5279621GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-796-21GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-796-21GM Supplier MilliporeSigma™ Supplier No. 5279621GM

missing translation for 'validMainframeMsg'

Antifungal polyene macrolide that is active in the cell membrane.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15-30°C
Purity or Quality Grade ≥80% by HPLC
Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.