Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Pirenzepine Dihydrochloride

Catalog No. 5057720001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-577-20001 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-577-20001 Supplier MilliporeSigma™ Supplier No. 5057720001
Add to Cart
Add to Cart

An antagonist selective for mAChR subtype M1.

An antagonist selective for mAChR subtype M1. Used for studying the functional roles of M1 receptors in both central and peripheral nervous systems.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 99% by HPLC
Target Proteins M1
Molecular Formula C19H21N5O2·2HCl
Solubility H2O
Content And Storage Protect from light
Form Solid
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.