Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PK11195

Catalog No. 5059810001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-598-10001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-598-10001 Supplier MilliporeSigma™ Supplier No. 5059810001
Add to Cart
Add to Cart

An isoquinoline carboxamide that acts as a potent ligand for peripheral bezodiazepine receptor (Kd = 20

An isoquinoline carboxamide that acts as a potent ligand for peripheral bezodiazepine receptor (Kd = 20nM). Exhibits anti-inflammatory properties. Binds to monocytes and following lesioning its binding up-regulated in brain macrophages and activated microglia. Shown to enhance the susceptibility of cells to apoptosis induction by DNA damaging agents and can reverse the suppression of apoptosis by Bcl-2. Serves as a ligand of the outer membrane translocator protein and reduces mitochondrial transmembrane potential within 30 minutes of exposure (˜100μM).
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Target Proteins peripheral bezodiazepine receptor
Molecular Formula C21H21ClN2O
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.