Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PKC Inhibitor Set

Catalog No. 5395731SET Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-957-31SET 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-957-31SET Supplier MilliporeSigma™ Supplier No. 5395731SET
Add to Cart
Add to Cart

The PKC Inhibitor Set controls the biological activity of PKC. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Contains 250μg of Bisindolylmaleimide I (Cat. No. 203290), 50μg of Calphostin C, Cladosporium cladosporioides (Cat. No. 208725), 5 mg of Chelerythrine Chloride (Cat. No. 220285), 500μg of Gö 6976 (Cat. No. 365250), 500μg of Myristoylated Protein Kinase C Inhibitor 20-28, Cell-Permeable (Cat. No. 476480), and 1 mg of Ro-32-0432 (Cat. No. 557525). Supplied with a data sheet.
TRUSTED_SUSTAINABILITY

Specifications

Target Proteins PKC
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 1 Set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.