Learn More
MilliporeSigma™ Calbiochem™ PKCη Pseudosubstrate Inhibitor, Myristoylated
Shop All MilliporeSigma ProductsDescription
- Peptide Sequence: Myr-Thr-Arg-Lys-Arg-Gln-Arg-Ala-Met-Arg-Arg-Arg-Val-His-Gln-Ile-Asn-Gly-OH
- Cell Permeable
Specifications
Specifications
| Formulation | Supplied as a trifluoroacetate salt. |
| Description | 95% by HPLC |
| Product Type | Inhibitor |
| Target Proteins | PKCν |
| Molecular Formula | C101H185N41O23S |
| Formula Weight | 2373.9g/mol |
| Solubility | H2O |
| Content And Storage | Hygroscopic |
| Form | Lyophilized |
| Quantity | 500 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.