Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PKCζ/ι Pseudosubstrate Inhibitor

Catalog No. 539610500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-961-0500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-961-0500UG Supplier MilliporeSigma™ Supplier No. 539610500UG
Add to Cart
Add to Cart

The PKCζ/ι Pseudosubstrate Inhibitor controls the biological activity of PKCζ/ι. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Peptide Sequence: H-Ser-Ile-Tyr-Arg-Arg-Gly-Ala-Arg-Arg-Trp-Arg-Lys-Leu-OH
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a trifluoroacetate salt.
Description 95% by HPLC
Target Proteins PKCζ
Molecular Formula C76H128N30O16
Formula Weight 1718g/mol
Solubility H2O or Glacial Acetic Acid
Content And Storage Hygroscopic
Form Lyophilized
Quantity 500 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.