Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PKG Inhibitor

Catalog No. 3706541MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-065-41MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-065-41MG Supplier MilliporeSigma™ Supplier No. 3706541MG
Add to Cart
Add to Cart

The PKG Inhibitor controls the biological activity of PKG. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Peptide Sequence: H-Arg-Lys-Arg-Ala-Arg-Lys-Glu-OH
TRUSTED_SUSTAINABILITY

Specifications

Description 95% by HPLC
Target Proteins PKG
Molecular Formula C38H74N18O10
Formula Weight 943.1g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Lyophilized
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.