Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
A cell-permeable quinoline-sulfonamide that acts as a potent allosteric PKM2 activator both in cell-free enzymatic assays (AC50 = 17nM with 40 ng PKM2/200μL) and in cultures (AC50 = 45nM in A549 cells) via a high affinity, 2:1 stoichiometric binding, effectively locking PKM2 in an active tetrameric state resistant to known intracellular negative regulators of FBP-activated PKM2 tetramer. PKM2 stimulation by compound treatment is shown to result in decreased serine biosynthesis (by 56%; 500nM for 24 h) with concomitant increase in serine influx as a compensating mechanism for maintaining cellular serine level necessary for supporting A549 proliferation. Simultaneous PKM2 activation and culture serine withdrawal results in cytostatic A549 growth arrest, but not apoptosis.
Specifications
Specifications
| Color | Off-white |
| Description | 99% by HPLC |
| Target Proteins | PKM2 |
| Molecular Formula | C27H26N4O4S |
| Solubility | DMSO |
| Content And Storage | Protect from light |
| Form | Powder |
| Quantity | 10 mg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?