Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Plasminogen Activator Inhibitor-1, Mutant, Human, Recombinant

Catalog No. 52820850UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-820-850UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-820-850UG Supplier MilliporeSigma™ Supplier No. 52820850UG
Add to Cart
Add to Cart

Primarily used for Protease Inhibitors applications.

Highly purified preparation of an altered form of human PAI-1 containing four mutated amino acids. Mutant PAI-1 is virtually unable to go latent and is stable at elevated temperature and pH for extended periods of time (t½ = 145 hours at 37°C, pH 7.4). Inhibits uPA (Ki = 5.1 x 106 M-1 sec-1) and tPA (Ki = 7.9 x 105 M-1 sec-1).
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 100 mM NaCl, 50 mM sodium phosphate buffer, 1 mM EDTA, pH 6.6.
Description 95% by SDS-PAGE assay
Target Proteins Plasminogen Activator
Formula Weight 43000g/mol
Content And Storage Avoid Freeze-Thaw Cycles
Form Liquid
Quantity 50 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.