Learn More
MilliporeSigma™ Calbiochem™ Plasminogen Activator Inhibitor-1, Mutant, Mouse, Recombinant
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Formulation | In 100 mM NaCl, 50 mM sodium acetate buffer, 1 mM EDTA, pH 5.0. |
| Description | 95% by SDS-PAGE |
| Target Proteins | Plasminogen Activator |
| Formula Weight | 43000g/mol |
| Content And Storage | Avoid Freeze-Thaw Cycles |
| Form | Liquid |
| Quantity | 50 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.