Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Polo-like Kinase Inhibitor II, BTO-1

Catalog No. 5282832MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-828-32MG 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-828-32MG Supplier MilliporeSigma™ Supplier No. 5282832MG
Add to Cart
Add to Cart

The Polo-like Kinase Inhibitor II, BTO-1, also referenced under CAS 40647-02-7, controls the biological activity of Polo-like Kinase. This small molecule/inhibitor is primarily used for Phosphorylation and Dephosphorylation applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Beige
Description 97% by HPLC
CAS 40647-02-7
Molecular Formula C9H4N4O4S
Formula Weight 264.2g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 2 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.