Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Porcn Inhibitor II, C59

Catalog No. 5004960001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-049-60001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-049-60001 Supplier MilliporeSigma™ Supplier No. 5004960001
Add to Cart
Add to Cart

A cell-permeable pyridinylphenyl-acetamide that is much more potent than IWP-2 (Cat

A cell-permeable pyridinylphenyl-acetamide that is much more potent than IWP-2 (Cat. No. 681671) in inhibiting MBOAT family member Porcupine-mediated Wnt palmitoylation (IC50 = 74 pM in STF reporter assays using Wnt3a-transfected HEK293). Shown to be effective against Porcn of murine and human, but not xenopus, species and potently inhibit the processing of both canonical and non-canonical Wnt subtypes. C59 is demonstrated to be orally active and effectively suppress MMTV-WNT1 tumor expansion in mice (5mg/kg and 10mg/kg p.o.) in vivo .

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 99% by HPLC
Target Proteins Porcupine (Porcn)-mediated Wnt palmitoylation
Molecular Formula C25H21N3O
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.