Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PP1 Analog V, 2,3-DMB-PP1

Catalog No. 52959910MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-959-910MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-959-910MG Supplier MilliporeSigma™ Supplier No. 52959910MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A cell-permeable 3-MB-PP1 analog that is more effective than 3-MB-PP1 in inhibiting Syk

A cell-permeable 3-MB-PP1 (Cat. No. 529582) analog that is more effective than 3-MB-PP1 in inhibiting SykM442A S505A ASKA- (analog-sensitive kinase alleles) mediated cellular calcium flux (98% vs. 20% inhibition, respectively, by 1μM 2,3-DMB-PP1 or 3-MB-PP1) in Syk MASA-expressing DT40 cells. Also shown to selectively inhibit L159G ASKA, but not wt, PDK1-dependent GSK3andalpha Ser21 and S6 Ser235/236 phosphorylation upon IGF1 stimulation in mES cells.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 97% by HPLC
CAS 956026-24-7
Molecular Formula C18H23N5
Formula Weight 309.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.