Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PP2A Activating Ligand, ITH12246

Catalog No. 5060740001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-607-40001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-607-40001 Supplier MilliporeSigma™ Supplier No. 5060740001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A cell-permeable, relatively non-toxic, 1,8-naphthyridine derived compound that blocks the inhibitory effect of protein phosphatase inhibitors, such as okadaic acid, on protein phosphatase 2A (PP2A)

A cell-permeable, relatively non-toxic, 1,8-naphthyridine derived compound that blocks the inhibitory effect of protein phosphatase inhibitors, such as okadaic acid, on protein phosphatase 2A (PP2A). Exhibits blood-brain barrier permeability. Protects neurons against β-amyloid peptides (Ab1-42) toxicity and okadaic acid (Cat. No. 459620)-induced tau hyperphosphorylation. Also protects against rotenone and oligomycin A induced neurotoxicity in SH-SY5Y neuroblastoma cells (at ∽300nM). Prevents the development of glutamate-induced neuronal lesions in rat hippocampal slices by up-regulating PP2A (∽ 3.0μM). Shown to reverse the scopolamine-induced memory loss in mice (∽10mg/kg i.p) and significantly reduces the infarct volume in an animal model of stroke (∽2.5mg/kg). Also acts as an inhibitor of acetylcholinesterase activity in Electrophorus electricus (IC50 = 60nM) and human erythrocytes (IC50 = 780nM).
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 98% by HPLC
Target Proteins PP2A
Molecular Formula C16H19N3O2·HCl
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.