Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Pravastatin, Sodium Salt

Catalog No. 52440325MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-440-325MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-440-325MG Supplier MilliporeSigma™ Supplier No. 52440325MG
Add to Cart
Add to Cart

A water-soluble, competitive 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase inhibitor that potently blocks in vivo cholesterol synthesis (Ki = ≈1 nM).

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Target Proteins AMG-CoA reductase
CAS 81131-70-6
Molecular Formula C23H35O7·Na
Formula Weight 446.5g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Crystalline Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.