Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PrCP Inhibitor

Catalog No. 5040440001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-404-40001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-404-40001 Supplier MilliporeSigma™ Supplier No. 5040440001
Add to Cart
Add to Cart

A dichlorobenzimidazolopyrrolidinamide compound

A dichlorobenzimidazolopyrrolidinamide compound that acts as a highly potent PrCP-selective inhibitor (IC50 = 1nM against human PrCP; IC50 = 2 and 8nM against mouse PrCP, respectively, in the absence or presence of 1% mouse serum albumin), presumably via non-covalent catalytic site interaction, while exhibiting no significant potency toward panels of ion channels, receptors, and enzymes, including homologous serine proteases QPP, FAP, PEP, ACE2, DPP4, DPP8, and DPP9 (IC50 >25μM). Oral administration (100mg/kg/d for 5 d) is reported to result in significant food intake reduction and body fat loss among high fat diet-induced obese mice in vivo .
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 97% by HPLC
Target Proteins PrCP
Molecular Formula C31H33Cl2N5O2
Solubility DMSO
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.