Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Prealbumin, Human Plasma

Catalog No. 5295771MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-957-71MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-957-71MG Supplier MilliporeSigma™ Supplier No. 5295771MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Transports retinol-binding protein, thyroxine, and vitamin A

  • Plasma protein with minimal carbohydrate
  • Found in plasma at about 30mg/100mL
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 150mM NaCl, 50mM sodium phosphate buffer, pH 7.5
Quantity 1 mg
Source Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.
Storage Requirements -20°C
Form Lyophilized
Purity or Quality Grade >95% by SDS-PAGE

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.