Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PRIMA-1

Catalog No. 53005010MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-005-010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-005-010MG Supplier MilliporeSigma™ Supplier No. 53005010MG
Add to Cart
Add to Cart

A cell-permeable quinuclidinone analog with antitumor properties.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 95% by NMR
Target Proteins Restores biochemical and biological function to mutant p53
CAS 5608-24-2
Molecular Formula C9H15NO3
Formula Weight 185.2g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.