Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Pristimerin

Catalog No. 5300705MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-007-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-007-05MG Supplier MilliporeSigma™ Supplier No. 5300705MG
Add to Cart
Add to Cart

A cell-permeable plant dienone-phenolic triterpenoid, naturally isolated from tripterygium wilfordii.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Description 95% by HPLC
CAS 1258-84-0
Molecular Formula C30H40O4
Formula Weight 464.6g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.