Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Pro-survival Compound, DDD00033325

Catalog No. 52965910MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-965-910MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-965-910MG Supplier MilliporeSigma™ Supplier No. 52965910MG
Add to Cart
Add to Cart

A cell-permeable, pyrrolidine small molecule that promotes single human embryonic stem cell survival up to 20-fold over background with equal or higher potency than Y-27632.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 95% by HPLC
Molecular Formula C17H19N5
Formula Weight 293.37g/mol
Solubility DMSO
Content And Storage Protect from light
Form Syrup
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.