Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ ProRS Inhibitor, Halofuginone

Catalog No. 5057630001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-576-30001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-576-30001 Supplier MilliporeSigma™ Supplier No. 5057630001
Add to Cart
Add to Cart

A cell-permeable racemic mixture of Halofuginone that inhibits of prolyl-tRNA synthetase.

  • Packaged Under Inert Gas
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Target Proteins prolyl-tRNA synthetase
CAS 64924-67-0
Molecular Formula C16H17BrClN3O3·HBr
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.