Learn More
MilliporeSigma™ Calbiochem™ Protease Inhibitor Cocktail Set VII
Catalog No. 5391381SET Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Set
Catalog No. Quantity
53-913-81SET 1 Set
1 options
Catalog No. 53-913-81SET Supplier MilliporeSigma™ Supplier No. 5391381SET
Add to Cart
Add to Cart
Protease Inhibitor Cocktail Set VII contains five protease inhibitors with broad specificity. Recommended for purification of proteins containing His·Tag™ sequences.
A cocktail of five protease inhibitors with broad specificity for the inhibition of cysteine, serine, aspartic, and thermolysin-like proteases and aminopeptidases. This cocktail is recommended for purification of proteins containing His Tag sequences. Each vial contains 100mm AEBSF, HCl (Cat. No. 101500), 5mm Bestatin (Cat. No. 200484), 1.5mm E-64 (Cat. No. 324890), 2mm Pepstatin A (Cat. No. 516481), and 200μM of Phosphoramidon, Disodium Salt (Cat. No. 525276). OnemL is recommended for the inhibition of proteases in 10 g of cells. Supplied with a data sheet. Note: 1 set = 5 x 1 ml.
TRUSTED_SUSTAINABILITY
Specifications
Formulation In 1mL DMSO.
Quantity 1 Set