Learn More
MilliporeSigma™ Calbiochem™ Protease Inhibitor Cocktail Set VIII
Catalog No. 5391291ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
53-912-91ML 1 mL
1 options
Catalog No. 53-912-91ML Supplier MilliporeSigma™ Supplier No. 5391291ML
Add to Cart
Add to Cart
The Protease Inhibitor Cocktail Set VIII controls the activity of Protease. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.
A cocktail of three protease inhibitors with selective specificity for the inhibition of cysteine proteases, including calpains, cathepsins, and papain. Each vial contains 1.56mm ALLN (Cat. No. 208719), 1.5mm E64 Protease Inhibitor (Cat. No. 324890), and 0.5mm Cathepsin Inhibitor I (Cat. No. 219415). Supplied with a data sheet. Note: 1 set = 5 x 1 ml.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Protease Inhibitor Cocktail
Form Liquid
Quantity 1 mL