Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Protease Inhibitor Cocktail Set X, EDTA-Free

Catalog No. 5391961SET Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-919-61SET 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-919-61SET Supplier MilliporeSigma™ Supplier No. 5391961SET
Add to Cart
Add to Cart

A cocktail of five protease inhibitors for the inhibition of a broad range of proteases, but not metalloproteases

A cocktail of five protease inhibitors for the inhibition of a broad range of proteases, but not metalloproteases. This cocktail is recommended for use with bacterial cells, animal tissues and plant extracts. Each vial, when reconstituted with 1mL H2O, yields a 100X stock solution. When diluted to 1X the cocktail contains 100μM AEBSF, HCl (Cat. No. 101500), 1μM Leupeptin Hemisulfate (Cat. No. 108975), 400μM Benzamidine, HCl (Cat. No. 199001), 1μM E-64 (Cat. No. 324890) and 150nM Aprotinin (Cat. No. 616370). Available as as a set of 5 x 1 ml.
TRUSTED_SUSTAINABILITY

Specifications

Target Proteins Proteases
Solubility H2O
Content And Storage Hygroscopic, Protect from Light, Protect from Moisture
Form Lyophilized
Quantity 1 Set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.